Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coro1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011779.3 | NP_035909.2 | 7 | 875 | ||
GenBank mRNA | AF143957.1 | - | 7 | 856 | ||
AK077552.1 | - | 7 | 885 | |||
AK079327.1 | - | 7 | 885 | |||
AK082986.1 | - | 7 | 877 | |||
AK088101.1 | - | 7 | 877 | |||
AK151017.1 | - | 7 | 864 | |||
AK151749.1 | - | 7 | 864 | |||
AK159506.1 | - | 7 | 867 | |||
AK167089.1 | - | 7 | 864 | |||
AK169262.1 | - | 7 | 860 | |||
AK169404.1 | - | 7 | 871 | |||
AK191781.1 | - | 2 | 82 | |||
BC009126.1 | - | 5 | 572 | |||
BC019444.1 | - | 7 | 846 | 4167810 | ||
BC085498.1 | - | 7 | 845 | 5716526 | ||
BC087553.1 | - | 7 | 863 | 30460345 | ||
BC099671.1 | - | 7 | 811 | 4918161 |