Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011785.3 | NP_035915.3 | 3 | 293 | ||
GenBank mRNA | AF124142.1 | - | 3 | 283 | ||
BC066861.1 | - | 3 | 293 | 30089997 | ||
CK390299.1 | - | 4 | 462 |