Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011789.2 | NP_035919.2 | 1 | 115 | ||
GenBank mRNA | AK140637.1 | - | 1 | 1088 | ||
KF481611.1 | - | 2 | 230 |