Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arih2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011790.4 | NP_035920.1 | 16 | 2163 | ||
GenBank mRNA | AJ130975.1 | - | 15 | 2023 | ||
AK011094.1 | - | 16 | 2170 | |||
AK045012.1 | - | 3484 | ||||
AK088772.1 | - | 1 | 1889 | |||
AK149936.1 | - | 16 | 2143 | |||
AK167104.1 | - | 16 | 2121 | |||
AK168055.1 | - | 16 | 2166 | |||
BC051998.1 | - | 16 | 2163 | 6400287 | ||
BC052422.1 | - | 16 | 2163 | 6400287 |