Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ets2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011809.3 | NP_035939.3 | 7 | 1025 | ||
GenBank mRNA | AK014602.2 | - | 7 | 947 | ||
AK030537.1 | - | 3 | 327 | |||
AK038348.1 | - | 7 | 1012 | |||
AK128933.1 | - | 8 | 899 | |||
AK143687.1 | - | 7 | 977 | |||
AK144974.1 | - | 7 | 976 | |||
AK145852.1 | - | 7 | 861 | |||
AK150175.1 | - | 7 | 954 | |||
AK152750.1 | - | 7 | 979 | |||
AK153339.1 | - | 7 | 977 | |||
AK153478.1 | - | 7 | 977 | |||
BC005486.1 | - | 7 | 946 | 3486021 | ||
BC005504.1 | - | 7 | 886 | 3490813 | ||
CT010235.1 | - | 6 | 787 | |||
J04103.1 | - | 7 | 924 |