Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fiz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110328.1 | NP_001103798.1 | 3 | 2434 | ||
NM_001110329.1 | NP_001103799.1 | 3 | 2377 | |||
NM_001110330.1 | NP_001103800.1 | 3 | 2307 | |||
NM_011813.3 | NP_035943.3 | 3 | 2418 | |||
GenBank mRNA | AF126746.1 | - | 3 | 2368 | ||
AF126747.1 | - | 3 | 2376 | |||
AK003641.1 | - | 1 | 1528 | |||
AK142205.1 | - | 3 | 2327 | |||
AK162454.1 | - | 3 | 2299 | |||
AK170633.1 | - | 3 | 2303 | |||
AK209331.1 | - | 1 | 180 | |||
AK220079.1 | - | 1 | 169 | |||
AK220212.1 | - | 3 | 2307 | |||
BC006633.1 | - | 3 | 2418 | 3482191 | ||
BC106113.1 | - | 3 | 2293 | 6492060 |