Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fyb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278269.1 | NP_001265198.1 | 1 | 267 | ||
NM_011815.5 | NP_035945.1 | 1 | 267 | |||
GenBank mRNA | AF001863.1 | - | 1 | 243 | ||
AF061744.1 | - | 1 | 243 | |||
AK030905.1 | - | 1 | 224 | |||
AK037454.1 | - | 1 | 268 | |||
AK038225.1 | - | 1 | 249 | |||
AK137907.1 | - | 1 | 222 | |||
AK172013.1 | - | 1 | 223 | |||
BC064102.1 | - | 1 | 203 | |||
BC141352.1 | - | 1 | 208 | 9055964 |