Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fyb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011815.5 | NP_035945.1 | 11 | 2087 | ||
GenBank mRNA | AF061744.1 | - | 11 | 2063 | ||
AK046759.1 | - | 2 | 1231 | |||
AK172164.1 | - | 4 | 319 | |||
BC141352.1 | - | 11 | 2028 | 9055964 |