Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gadd45g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011817.2 | NP_035947.2 | 3 | 377 | ||
GenBank mRNA | AB021884.1 | - | 3 | 374 | ||
AF055638.1 | - | 3 | 365 | |||
AK002237.1 | - | 3 | 375 | |||
AK007410.1 | - | 3 | 380 | |||
BC001989.1 | - | 3 | 358 | 3493618 |