Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hax1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282032.1 | NP_001268961.1 | 6 | 850 | ||
NM_011826.4 | NP_035956.1 | 7 | 1102 | |||
GenBank mRNA | AF023482.1 | - | 7 | 1074 | ||
AF023483.1 | - | 6 | 1237 | |||
AK002256.1 | - | 7 | 1053 | |||
AK010633.1 | - | 7 | 1086 | |||
AK088746.1 | - | 7 | 1084 | |||
AK135138.1 | - | 6 | 812 | |||
AK203815.1 | - | 1 | 144 | |||
BC006688.1 | - | 7 | 1031 | 3582652 | ||
BC098225.1 | - | 7 | 1056 | 1494110 |
Gene Symbol: Gm14270 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK198525.1 | - | 1 | 145 |