Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMEL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200054.1 | NP_001186983.1 | 2 | 169 | ||
NM_001320121.1 | NP_001307050.1 | 2 | 169 | |||
NM_001320122.1 | NP_001307051.1 | 2 | 169 | |||
NM_006928.4 | NP_008859.1 | 3 | 204 | |||
XM_006719569.1 | XP_006719632.1 | 2 | 781 | |||
XM_011538685.1 | XP_011536987.1 | 3 | 233 | |||
GenBank mRNA | AK092881.1 | - | 2 | 782 | ||
AK130180.1 | - | 3 | 228 | |||
BC001414.2 | - | 2 | 128 | 3139788 | ||
BT007202.1 | - | 2 | 118 | |||
DA035117.1 | - | 3 | 201 | |||
DA633173.1 | - | 3 | 195 | |||
M32295.1 | - | 2 | 139 | |||
M77348.1 | - | 2 | 129 | |||
S73003.1 | - | 2 | 139 | |||
U01874.1 | - | 2 | 124 |