Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nek4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF099067.1 | - | 15 | 2484 | ||
AJ223071.1 | - | 15 | 2574 | |||
AK078809.1 | - | 14 | 2451 | |||
AK132803.1 | - | 14 | 2368 | |||
AK147381.1 | - | 12 | 6271 | |||
AK149147.1 | - | 6 | 652 | |||
AK149584.1 | - | 14 | 2368 | |||
BC057939.1 | - | 15 | 2486 | 5325558 | ||
Y09234.1 | - | 14 | 2430 |