Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tenm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011858.3 | NP_035988.2 | 4, 5 | 585 | ||
GenBank mRNA | AB025413.1 | - | 2, 3 | 418 | ||
AF059485.1 | - | 5, 6 | 813 | |||
AK147329.1 | - | 4, 5 | 586 | |||
AK147579.1 | - | 4, 5 | 774 |