Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024644.1 | NP_001019815.1 | 2 | 890 | ||
NM_005283.2 | NP_005274.1 | 3 | 1012 | |||
XM_017006196.1 | XP_016861685.1 | 4 | 2030 | |||
XM_017006197.1 | XP_016861686.1 | 6 | 2282 | |||
XM_017006198.1 | XP_016861687.1 | 5 | 2095 | |||
XM_017006199.1 | XP_016861688.1 | 4 | 1092 | |||
XM_017006200.1 | XP_016861689.1 | 3 | 1950 | |||
XM_017006201.1 | XP_016861690.1 | 2 | 1799 | |||
GenBank mRNA | BC069075.1 | - | 1 | 788 | 7216889 | |
BC069589.1 | - | 1 | 788 | 7262280 | ||
BC095533.1 | - | 1 | 839 | 6971949 |