Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dazap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011873.2 | NP_036003.2 | 2, 3 | 190 | ||
GenBank mRNA | AF085348.1 | - | 2, 3 | 155 | ||
AK002595.1 | - | 2, 3 | 227 | |||
AK050159.1 | - | 2, 3 | 204 | |||
AK088916.1 | - | 2, 3 | 212 | |||
AK132139.1 | - | 2, 3 | 204 | |||
AK137246.1 | - | 2, 3 | 191 | |||
AK145398.1 | - | 2, 3 | 224 | |||
AK146970.1 | - | 2, 3 | 215 | |||
BC014808.1 | - | 2, 3 | 190 | 4506124 | ||
BC029351.1 | - | 1, 2 | 121 | |||
BC083347.1 | - | 2, 3 | 182 | 6829350 |