Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dazap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011873.2 | NP_036003.2 | 4 | 1586 | ||
GenBank mRNA | AF085348.1 | - | 4 | 1550 | ||
AK002595.1 | - | 4 | 1623 | |||
AK050159.1 | - | 4 | 1600 | |||
AK088916.1 | - | 4 | 1608 | |||
AK132139.1 | - | 4 | 1600 | |||
AK137246.1 | - | 4 | 1587 | |||
AK145398.1 | - | 4 | 1620 | |||
AK146970.1 | - | 4 | 1611 | |||
BC014808.1 | - | 4 | 1586 | 4506124 | ||
BC029351.1 | - | 3 | 1517 | |||
BC083347.1 | - | 4 | 1578 | 6829350 |