Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011874.2 | NP_036004.2 | 5, 6 | 658 | ||
GenBank mRNA | AK050677.1 | - | 5, 6 | 636 | ||
AK077507.1 | - | 5, 6 | 621 | |||
AK150989.1 | - | 5, 6 | 613 | |||
AK153991.1 | - | 5, 6 | 641 | |||
AK160644.1 | - | 5, 6 | 620 | |||
AK160718.1 | - | 5, 6 | 612 | |||
AK167041.1 | - | 5, 6 | 603 | |||
AK167528.1 | - | 5, 6 | 613 | |||
AK169224.1 | - | 5, 6 | 621 | |||
AK169244.1 | - | 5, 6 | 1121 | |||
BC012708.1 | - | 5, 6 | 619 | 4211677 | ||
BC092265.1 | - | 5, 6 | 602 | 3671250 | ||
L76223.1 | - | 5, 6 | 609 |