Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPTBN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006946.2 | NP_008877.1 | 2 | 214 | ||
XM_005274192.4 | XP_005274249.1 | 3 | 876 | |||
XM_005274193.3 | XP_005274250.1 | 2 | 650 | |||
XM_006718671.3 | XP_006718734.1 | 3 | 276 | |||
XM_017018174.1 | XP_016873663.1 | 3 | 968 | |||
XM_017018175.1 | XP_016873664.1 | 3 | 597 | |||
XM_017018176.1 | XP_016873665.1 | 3 | 317 | |||
XM_017018177.1 | XP_016873666.1 | 2 | 506 | |||
XM_017018178.1 | XP_016873667.1 | 2 | 189 | |||
GenBank mRNA | AB008567.1 | - | 2 | 214 | ||
BC139788.1 | - | 2 | 185 |