Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF037205.1 | - | 4 | 313 | ||
AF037206.1 | - | 4 | 313 | |||
AI882163.1 | - | 4 | 372 | |||
AK034135.1 | - | 4 | 364 | |||
AK036591.1 | - | 4 | 358 | |||
AK050095.1 | - | 4 | 326 | |||
AK082296.1 | - | 4 | 354 | |||
AK139447.1 | - | 4 | 319 | |||
AK140682.1 | - | 4 | 368 | |||
AK158046.1 | - | 4 | 354 | |||
AK167278.1 | - | 4 | 318 | |||
BB847350.1 | - | 4 | 379 | |||
BC058182.1 | - | 4 | 321 | 5324040 |