Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rngtt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF025653.1 | - | 2 | 168 | ||
AF034568.1 | - | 2 | 222 | |||
AK002922.1 | - | 2 | 271 | |||
AK079488.1 | - | 2 | 268 | |||
AK142016.1 | - | 2 | 265 | |||
AK150497.1 | - | 2 | 275 | |||
AK151463.1 | - | 2 | 289 | |||
AK160007.1 | - | 2 | 277 | |||
AK160706.1 | - | 2 | 255 | |||
AK169305.1 | - | 2 | 241 | |||
BC043657.1 | - | 2 | 240 | 5343539 |