Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rngtt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF025653.1 | - | 11 | 1195 | ||
AF034568.1 | - | 11 | 1249 | |||
AK002922.1 | - | 11 | 1298 | |||
AK142016.1 | - | 11 | 1292 | |||
AK150497.1 | - | 11 | 1302 | |||
AK151463.1 | - | 11 | 1316 | |||
AK160007.1 | - | 11 | 1304 | |||
AK160706.1 | - | 11 | 1282 | |||
AK169305.1 | - | 11 | 1268 | |||
BC043657.1 | - | 11 | 1267 | 5343539 |