Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011891.4 | NP_036021.1 | 3 | 316 | ||
GenBank mRNA | AB024923.1 | - | 3 | 537 | ||
AK040974.1 | - | 3 | 318 | |||
BC130019.1 | - | 3 | 306 | 40140624 |