Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145858.1 | NP_001139330.1 | 5 | 638 | ||
NM_001145859.1 | NP_001139331.1 | 5 | 641 | |||
NM_011893.3 | NP_036023.3 | 5 | 538 | |||
GenBank mRNA | AK089455.1 | - | 5 | 469 | ||
AK143548.1 | - | 5 | 643 | |||
AK150265.1 | - | 5 | 640 | |||
AK155103.1 | - | 5 | 495 | |||
AK155898.1 | - | 5 | 538 | |||
AK155970.1 | - | 5 | 586 | |||
AK156912.1 | - | 5 | 583 | |||
BC010198.1 | - | 5 | 476 | 3967543 | ||
BC085497.1 | - | 5 | 529 | 5696296 | ||
L14543.1 | - | 5 | 472 |