Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3bp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011894.2 | NP_036024.2 | 3 | 335 | ||
GenBank mRNA | AB016835.1 | - | 3 | 163 | ||
AK043375.1 | - | 380 | ||||
AK161427.1 | - | 2 | 317 | |||
AK165033.1 | - | 3 | 369 | |||
BC018237.1 | - | 3 | 367 | 3482670 | ||
BC053741.1 | - | 3 | 335 | 6336511 |