Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3bp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011894.2 | NP_036024.2 | 6 | 738 | ||
GenBank mRNA | AB016835.1 | - | 6 | 566 | ||
AK043375.1 | - | 783 | ||||
AK161427.1 | - | 5 | 720 | |||
AK165033.1 | - | 6 | 772 | |||
BC018237.1 | - | 6 | 770 | 3482670 | ||
BC053741.1 | - | 6 | 738 | 6336511 |