Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpdu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301710.1 | NP_001288639.1 | 4 | 428 | ||
NM_001301711.1 | NP_001288640.1 | 6 | 668 | |||
NM_011900.4 | NP_036030.2 | 6 | 633 | |||
GenBank mRNA | AB024713.1 | - | 6 | 561 | ||
AK040432.1 | - | 6 | 572 | |||
AK080233.1 | - | 6 | 627 | |||
AK089426.1 | - | 6 | 575 | |||
AK172020.1 | - | 6 | 594 | |||
AK188244.1 | - | 1 | 24 | |||
BC026776.1 | - | 6 | 555 | 4159355 | ||
BC048871.1 | - | 6 | 561 | 5364408 | ||
CV561188.1 | - | 6 | 619 | |||
CV561394.1 | - | 5 | 467 | |||
W67006.1 | - | 2 | 135 |