Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112705.2 | NP_001106176.1 | 20 | 2435 | ||
NM_001294329.1 | NP_001281258.1 | 18 | 2295 | |||
NM_001294330.1 | NP_001281259.1 | 18 | 2243 | |||
NM_001294331.1 | NP_001281260.1 | 20 | 2448 | |||
NM_001294334.1 | NP_001281263.1 | 20 | 2435 | |||
GenBank mRNA | AF045252.1 | - | 20 | 2454 | ||
AF045253.1 | - | 19 | 1882 | |||
AF045254.1 | - | 15 | 1596 | |||
AK014829.1 | - | 20 | 2073 | |||
AK161383.1 | - | 2 | 203 | |||
AK165667.1 | - | 20 | 2438 | |||
AX775743.1 | - | 14 | 1460 | |||
BC066198.1 | - | 18 | 1788 | |||
U56909.1 | - | 11 | 1133 |