Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tll2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011904.3 | NP_036034.1 | 4 | 576 | ||
GenBank mRNA | AF073526.1 | - | 4 | 637 | ||
BC132537.1 | - | 4 | 576 | 40130814 | ||
BC145132.1 | - | 4 | 576 | 9053680 |