Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FES | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143783.1 | NP_001137255.1 | 2 | 233 | ||
NM_001143784.1 | NP_001137256.1 | 3 | 407 | |||
NM_001143785.1 | NP_001137257.1 | 3 | 352 | |||
NM_002005.3 | NP_001996.1 | 4 | 503 | |||
XM_005254880.1 | XP_005254937.1 | 3 | 318 | |||
XM_005254882.1 | XP_005254939.1 | 4 | 519 | |||
XM_017022005.1 | XP_016877494.1 | 4 | 487 | |||
XM_017022006.1 | XP_016877495.1 | 3 | 360 | |||
XM_017022007.1 | XP_016877496.1 | 4 | 487 | |||
XM_017022008.1 | XP_016877497.1 | 3 | 361 | |||
XM_017022009.1 | XP_016877498.1 | 4 | 584 | |||
XM_017022010.1 | XP_016877499.1 | 3 | 435 | |||
GenBank mRNA | AK300595.1 | - | 3 | 342 | ||
AK300605.1 | - | 4 | 461 | |||
AK309530.1 | - | 3 | 764 | |||
AK312545.1 | - | 4 | 492 | |||
AY513654.1 | - | 2 | 233 | |||
AY513655.1 | - | 2 | 233 | |||
AY513656.1 | - | 2 | 233 | |||
AY513657.1 | - | 3 | 407 | |||
BC035357.1 | - | 4 | 479 | 5170548 | ||
DA782386.1 | - | 4 | 526 | |||
DB264752.1 | - | 4 | 549 | |||
X52192.1 | - | 4 | 482 |