Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xrn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011917.2 | NP_036047.2 | 2 | 160 | ||
GenBank mRNA | AK004800.1 | - | 2 | 135 | ||
AK011711.1 | - | 2 | 158 | |||
AK031247.1 | - | 2 | 203 | |||
AK053643.1 | - | 2 | 153 | |||
AK133654.1 | - | 2 | 142 | |||
AK163899.1 | - | 2 | 138 | |||
AK167019.1 | - | 2 | 180 | |||
AK168037.1 | - | 2 | 206 | |||
BC052395.1 | - | 2 | 136 | |||
BC052823.1 | - | 2 | 125 | |||
BC054743.1 | - | 2 | 160 | 6400974 | ||
D38517.1 | - | 2 | 98 |