Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ing1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB221554.1 | - | 2 | 446 | ||
AF177755.1 | - | 2 | 428 | |||
AF177756.1 | - | 2 | 413 | |||
AF177757.1 | - | 2 | 1286 | |||
AK035490.1 | - | 2 | 443 | |||
AK076482.1 | - | 2 | 336 | |||
AK141466.1 | - | 2 | 336 | |||
AK145553.1 | - | 2 | 338 | |||
AK157424.1 | - | 2 | 435 | |||
BC016573.1 | - | 2 | 828 | |||
BC147770.1 | - | 2 | 538 | 9087493 | ||
BC147784.1 | - | 2 | 538 | 9087503 | ||
CK624100.1 | - | 2 | 324 | |||
DV655694.1 | - | 2 | 406 |