Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Angptl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011923.4 | NP_036053.2 | 3 | 1534 | ||
GenBank mRNA | AF125176.1 | - | 2 | 1021 | ||
AK037265.1 | - | 3 | 1427 | |||
AK155464.1 | - | 3 | 1535 | |||
BC138609.1 | - | 3 | 1340 | 8861629 | ||
BC138610.1 | - | 3 | 1340 | 8861630 |