Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dapp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011932.2 | NP_036062.1 | 4 | 544 | ||
GenBank mRNA | AF163255.1 | - | 4 | 498 | ||
AF186023.1 | - | 4 | 519 | |||
AK076886.1 | - | 2 | 97 | |||
AK145745.1 | - | 4 | 510 | |||
AK146180.1 | - | 4 | 513 | |||
AK169584.1 | - | 3 | 409 | |||
BC014759.1 | - | 4 | 540 | 4486834 |