Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map3k1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011945.2 | NP_036075.2 | 20 | 4497 | ||
GenBank mRNA | AF117340.1 | - | 20 | 4497 | ||
L13103.1 | - | 10 | 2506 | |||
U23470.1 | - | 7 | 2098 |