Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map3k2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011946.3 | NP_036076.2 | 5 | 710 | ||
GenBank mRNA | AK143367.1 | - | 5 | 707 | ||
U43186.1 | - | 5 | 608 |