Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168508.1 | NP_001161980.1 | 11 | 1310 | ||
NM_001168513.1 | NP_001161985.1 | 11 | 911 | |||
NM_001168514.1 | NP_001161986.1 | 12 | 1355 | |||
NM_011951.3 | NP_036081.1 | 11 | 1310 | |||
GenBank mRNA | AF195850.1 | - | 8 | 633 | ||
AK089059.1 | - | 11 | 1224 | |||
AK133684.1 | - | 11 | 1223 | |||
AK151348.1 | - | 11 | 912 | |||
AK153025.1 | - | 11 | 912 | |||
BC012235.1 | - | 11 | 1310 | 4195842 | ||
BC018227.1 | - | 12 | 1252 | |||
BC085303.1 | - | 11 | 1286 | 5389457 | ||
CX568915.1 | - | 10 | 704 | |||
D83073.1 | - | 11 | 1141 | |||
U10871.1 | - | 11 | 1208 |