Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011952.2 | NP_036082.1 | 9 | 1599 | ||
GenBank mRNA | AK155287.1 | - | 8 | 1541 | ||
BC013754.1 | - | 9 | 1432 | |||
BC029712.1 | - | 9 | 1593 | 5041517 | ||
Z14249.1 | - | 9 | 1569 |