Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nubp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011955.2 | NP_036085.1 | 3 | 202 | ||
GenBank mRNA | AF114170.1 | - | 3 | 179 | ||
AK020876.1 | - | 3 | 187 | |||
AK157212.1 | - | 3 | 194 | |||
BC031430.1 | - | 3 | 192 | |||
BC047926.1 | - | 3 | 180 | |||
BC055436.1 | - | 3 | 190 | 3660324 | ||
CJ183745.1 | - | 3 | 202 |