Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011960.2 | NP_036090.2 | 3 | 817 | ||
GenBank mRNA | AF079557.1 | - | 3 | 566 | ||
AK036656.1 | - | 3 | 840 | |||
AK080306.1 | - | 3 | 537 | |||
AK160742.1 | - | 2 | 291 | |||
AK164013.1 | - | 3 | 804 | |||
BC050892.1 | - | 3 | 816 | |||
BC053037.1 | - | 3 | 800 | |||
BC059827.1 | - | 3 | 826 |