Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011960.2 | NP_036090.2 | 3 | 1331 | ||
GenBank mRNA | AF079557.1 | - | 3 | 1080 | ||
AK036656.1 | - | 3 | 1354 | |||
AK160742.1 | - | 2 | 805 | |||
AK163388.1 | - | 3 | 1358 | |||
AK164013.1 | - | 3 | 1318 | |||
BC050892.1 | - | 3 | 1330 | |||
BC053037.1 | - | 3 | 1314 | |||
BC059827.1 | - | 3 | 1340 |