Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011990.2 | NP_036120.1 | 3 | 772 | ||
GenBank mRNA | AB022345.1 | - | 3 | 739 | ||
AK037490.1 | - | 3 | 773 | |||
AK037742.1 | - | 3 | 747 | |||
AK089202.1 | - | 3 | 745 | |||
AK162233.1 | - | 3 | 545 | |||
AK163168.1 | - | 3 | 746 | |||
AK165599.1 | - | 3 | 748 | |||
AK171192.1 | - | 3 | 770 | |||
AK171252.1 | - | 3 | 746 | |||
AY766236.1 | - | 3 | 739 | |||
AY766237.1 | - | 739 | ||||
BC141402.1 | - | 3 | 669 | 9056015 | ||
BC141408.1 | - | 3 | 669 | 9056021 |