Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011991.1 | NP_036121.1 | 7 | 701 | ||
GenBank mRNA | AF071313.1 | - | 7 | 701 | ||
AK048906.1 | - | 7 | 691 | |||
AK142318.1 | - | 7 | 687 | |||
BC068179.1 | - | 7 | 701 | 30633689 |