Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpysl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011993.4 | NP_036123.3 | 14 | 2427 | ||
GenBank mRNA | AB006715.1 | - | 14 | 2357 | ||
AK090197.1 | - | 13 | 2957 | |||
AK145638.1 | - | 13 | 2530 | |||
AK165694.1 | - | 14 | 2428 | |||
BC046333.1 | - | 13 | 3980 | |||
BC090674.1 | - | 2399 |