Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011994.2 | NP_036124.2 | 2 | 1179 | ||
GenBank mRNA | AK051445.1 | - | 2 | 1153 | ||
AK082588.1 | - | 2 | 1151 | |||
AK087653.1 | - | 2 | 798 | |||
AK161027.1 | - | 2 | 840 | |||
BC019187.1 | - | 2 | 1123 | 5027149 | ||
Z48670.1 | - | 2 | 1183 |