Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cln8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012000.3 | NP_036130.1 | 2 | 839 | ||
GenBank mRNA | AF125307.1 | - | 2 | 329 | ||
AF125308.1 | - | 2 | 329 | |||
AK049145.1 | - | 2 | 512 | |||
AK080049.1 | - | 2 | 347 | |||
AK088300.1 | - | 2 | 493 | |||
AK089692.1 | - | 2 | 488 | |||
AK128890.1 | - | 2 | 510 | |||
AK164268.1 | - | 2 | 512 | |||
AK166905.1 | - | 2 | 1264 | |||
AK169698.1 | - | 2 | 493 | |||
AK171446.1 | - | 2 | 521 | |||
BC021625.1 | - | 2 | 345 | 4188527 |