Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012002.3 | NP_036132.1 | 4, 5 | 433 | ||
GenBank mRNA | AF071315.1 | - | 4, 5 | 470 | ||
AK138409.1 | - | 3, 4 | 559 | |||
AK146410.1 | - | 4, 5 | 444 | |||
AK146775.1 | - | 4, 5 | 446 | |||
BC004664.1 | - | 4, 5 | 432 | 3498214 | ||
BC014286.1 | - | 4, 5 | 437 | 4163277 |