Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx3y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012008.2 | NP_036138.1 | 4 | 336 | ||
GenBank mRNA | AJ007376.1 | - | 4 | 303 | ||
AK154382.1 | - | 4 | 296 | |||
BC021453.1 | - | 4 | 301 | 5043470 | ||
BC051046.1 | - | 4 | 325 | |||
CD552910.1 | - | 4 | 336 |