Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx3y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012008.2 | NP_036138.1 | 13 | 1563 | ||
GenBank mRNA | AJ007376.1 | - | 13 | 1530 | ||
AK154382.1 | - | 13 | 1523 | |||
BC021453.1 | - | 13 | 1528 | 5043470 | ||
BC051046.1 | - | 13 | 1552 | |||
L25337.1 | - | 5 | 628 |