Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx3y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012008.2 | NP_036138.1 | 17 | 3052 | ||
GenBank mRNA | AJ007376.1 | - | 17 | 3017 | ||
AK154382.1 | - | 17 | 3012 | |||
AK160604.1 | - | 4 | 1538 | |||
BC021453.1 | - | 17 | 3017 | 5043470 |