Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2s3x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012010.3 | NP_036140.1 | 4 | 390 | ||
GenBank mRNA | AJ006587.1 | - | 4 | 380 | ||
AK010819.1 | - | 1 | 68 | |||
AK128949.1 | - | 4 | 389 | |||
AK146900.1 | - | 4 | 431 | |||
AK150095.1 | - | 4 | 397 | |||
AK165866.1 | - | 4 | 392 | |||
AK171317.1 | - | 4 | 391 | |||
BC036993.1 | - | 4 | 388 | |||
BC063755.1 | - | 4 | 390 | 6513251 | ||
BC139234.1 | - | 4 | 390 | 8862256 | ||
BC139235.1 | - | 4 | 390 | 8862257 |